Merck
Association between in vitro susceptibility to natamycin and voriconazole and clinical outcomes in fungal keratitis.
Ophthalmology (2014-04-22)
Catherine Q Sun, Prajna Lalitha, N Venkatesh Prajna, Rajarathinam Karpagam, Manoharan Geetha, Kieran S O'Brien, Catherine E Oldenburg, Kathryn J Ray, Stephen D McLeod, Nisha R Acharya, Thomas M Lietman
RESUMEN
To assess the association between minimum inhibitory concentration (MIC) and clinical outcomes in a fungal keratitis clinical trial. Experimental study using data from a randomized comparative trial. Of the 323 patients enrolled in the trial, we were able to obtain MIC values from 221 patients with monocular fungal keratitis. The Mycotic Ulcer Treatment Trial I was a randomized, double-masked clinical trial comparing clinical outcomes of monotherapy with topical natamycin versus voriconazole for the treatment of fungal keratitis. Speciation and determination of MIC to natamycin and voriconazole were performed according to Clinical and Laboratory Standards Institute guidelines. The relationship between MIC and clinical outcome was assessed. The primary outcome was 3-month best spectacle-corrected visual acuity. Secondary outcomes included 3-month infiltrate or scar size; corneal perforation and/or therapeutic penetrating keratoplasty; and time to re-epithelialization. A 2-fold increase in MIC was associated with a larger 3-month infiltrate or scar size (0.21 mm; 95% confidence interval [CI], 0.10-0.31; P < 0.001) and increased odds of perforation (odds ratio, 1.32; 95% CI, 1.04-1.69; P = 0.02). No correlation was found between MIC and 3-month visual acuity. For natamycin-treated cases, an association was found between higher natamycin MIC with larger 3-month infiltrate or scar size (0.29 mm; 95% CI, 0.15-0.43; P < 0.001) and increased perforations (odds ratio, 2.41; 95% CI, 1.46-3.97; P < 0.001). Among voriconazole-treated cases, the voriconazole MIC did not correlate with any of the measured outcomes in the study. Decreased susceptibility to natamycin was associated with increased infiltrate or scar size and increased odds of perforation. There was no association between susceptibility to voriconazole and outcome.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Pimaricin, from Streptomyces chattanoogensis, ≥95% (HPLC)
Sigma-Aldrich
Voriconazole, ≥98% (HPLC)
Sigma-Aldrich
Pimaricin preparation, ~2.5% (γ-irradiated Pimaricin), aqueous suspension
Supelco
Natamycin, VETRANAL®, analytical standard
Supelco
Voriconazole, VETRANAL®, analytical standard
Voriconazole, European Pharmacopoeia (EP) Reference Standard