Merck
Ring cleavage and degradative pathway of cyanuric acid in bacteria.
The Biochemical journal (1985-10-01)
A M Cook, P Beilstein, H Grossenbacher, R Hütter
RESUMEN
The degradative pathway of cyanuric acid [1,3,5-triazine-2,4,6(1H,3H,5H)-trione] was examined in Pseudomonas sp. strain D. The bacterium grew with cyanuric acid, biuret, urea or NH4+ as sole source of nitrogen, and each substrate was entirely metabolized concomitantly with growth. Enzymes from strain D were separated by chromatography on DEAE-cellulose and three reactions were examined. Cyanuric acid (1 mol) was converted stoichiometrically into 1.0 mol of CO2 and 1.1 mol of biuret, which was conclusively identified. Biuret (1 mol) was converted stoichiometrically into 1.1 mol of NH4+, about 1 mol of CO2 and 1.0 mol of urea, which was conclusively identified. Urea (1 mol) was converted into 1.9 mol of NH4+ and 1.0 mol of CO2. The reactions proceeded under aerobic or anoxic conditions and were presumed to be hydrolytic. Data indicate that the same pathway occurred in another pseudomonad and a strain of Klebsiella pneumoniae.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cyanuric acid, 98%