Merck
Immobilised vitamin B12 as a biomimetic model for base-off/histidine-on coordination.
Chemical communications (Cambridge, England) (2011-09-20)
Christine Männel-Croisé, Felix Zelder
RESUMEN
Immobilisation of vitamin B12 allows synthesising a biomimetic base-off/histidine-on complex that resembles structural features of cobalamin dependent enzyme active sites.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
5,6-Dimethylbenzimidazole, ≥99%