Merck
Stereoselective syntheses of the antihistaminic drug olopatadine and its E-isomer.
The Journal of organic chemistry (2012-06-27)
Joan Bosch, Jordi Bachs, Antonia M Gómez, Rosa Griera, Marta Écija, Mercedes Amat
RESUMEN
Practical stereoselective synthetic routes to the antihistaminic drug olopatadine and its E-isomer have been developed, the key steps being a trans stereoselective Wittig olefination using a nonstabilized phosphorus ylide and a stereoselective Heck cyclization. The stereoselectivity of the Wittig reaction depends on both the phosphonium salt anion and the cation present in the base used to generate the ylide.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Olopatadine hydrochloride, ≥98% (HPLC)