- High-pressure glycosylations of unreactive alcohols and the formation of N-glycosyl collidinium salts.
High-pressure glycosylations of unreactive alcohols and the formation of N-glycosyl collidinium salts.
Carbohydrate research (1990-08-01)
W G Dauben, P Köhler
PMID2224903
RESUMEN
The yields of disaccharide glycosylation products in tetramethylammonium bromide or silver triflate-collidine activated reactions between hindered alcohols and glycosyl halides were not greatly ++affected when a pressure of 15 kbar was applied. The formation of orthoester products was greatly increased under pressure. When orthoester formation was not possible both disaccharides and the related N-glycosyl collidinium salts were found.