Merck
Lethal effects of abamectin on the aquatic organisms Daphnia similis, Chironomus xanthus and Danio rerio.
Chemosphere (2011-10-01)
Andréa Novelli, Bruna Horvath Vieira, Daniela Cordeiro, Luciana Teresa Dias Cappelini, Eny Maria Vieira, Evaldo Luiz Gaeta Espíndola
RESUMEN
Abamectin is used as an acaricide and insecticide for fruits, vegetables and ornamental plants, as well as a parasiticide for animals. One of the major problems of applying pesticides to crops is the likelihood of contaminating aquatic ecosystems by drift or runoff. Therefore, toxicity tests in the laboratory are important tools to predict the effects of chemical substances in aquatic ecosystems. The aim of this study was to assess the potential hazards of abamectin to the freshwater biota and consequently the possible losses of ecological services in contaminated water bodies. For this purpose, we identified the toxicity of abamectin on daphnids, insects and fish. Abamectin was highly toxic, with an EC(50) 48 h for Daphnia similis of 5.1 ng L(-1), LC(50) 96 h for Chironomus xanthus of 2.67 μg L(-1) and LC(50) 48 h for Danio rerio of 33 μg L(-1).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Abamectin, PESTANAL®, analytical standard