Merck
Fusion expression of cecropin B-like antibacterial peptide in Escherichia coli and preparation of its antiserum.
Biotechnology letters (2010-01-05)
Fuxian Yu, Jigui Wang, Peng Zhang, Yan Hong, Weiquan Liu
RESUMEN
Antibacterial peptides have a broad range of antibacterial properties that makes them highly toxic for expression in Escherichia coli. For prepare an antiserum to detect these peptides, we developed a cecropin B mutant with a green fluorescent protein fusion partner resulting in high expression of a 37 kDa fusion peptide in E. coli with a yield of 7.9 mg/l culture medium after purification on Ni-IDA resin. Guinea pigs when immunized with the fusion peptide produced a specific antiserum which titers in excess of 1:25,600.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cecropin B, ≥97% (HPLC), powder