Merck
  • An analysis of the cortical and striatal involvement in dyskinesia induced in rats by intracerebral injection of GABA-transaminase inhibitors and picrotoxin.
An analysis of the cortical and striatal involvement in dyskinesia induced in rats by intracerebral injection of GABA-transaminase inhibitors and picrotoxin.
European journal of pharmacology (1980-04-04)
M M Robin, M G Palfreyman, M M Zraika, P J Schechter
RESUMEN
Unilateral intrastriatal injection of various substances induces a characteristic dyskinetic syndrome in rats. These substances include picrotoxin as well as a series of irreversible GABA-transaminase inhibitors. Using the degree of enzyme inhibition in various brain areas as a measure of drug distribution following intrastriatal administration of gamma-acetylenic GABA and gamma-vinyl GABA, there was found considerable retrodiffusion via the needle tract to the overlying cortex. Topical application of gamma-acetylenic GABA and gamma-vinyl GABA to the cortical surface overlying the striatum produced a high incidence of identical dyskinesias without any evidence of diffusion of drugs to the striatum. The cortically induced movements could be duplicated by picrotoxin application to a defined cortical area. These findings suggest that interference with gabaergic function in the striatum is not necessary for the production of the dyskinetic syndrome and that this syndrome may be a cortically induced phenomenon.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
S(+)-γ-Vigabatrin, solid