Merck
Cortical dynein is critical for proper spindle positioning in human cells.
The Journal of cell biology (2012-10-03)
Sachin Kotak, Coralie Busso, Pierre Gönczy
RESUMEN
Correct spindle positioning is fundamental for proper cell division during development and in stem cell lineages. Dynein and an evolutionarily conserved ternary complex (nuclear mitotic apparatus protein [NuMA]-LGN-Gα in human cells and LIN-5-GPR-1/2-Gα in Caenorhabditis elegans) are required for correct spindle positioning, but their relationship remains incompletely understood. By analyzing fixed specimens and conducting live-imaging experiments, we uncovered that appropriate levels of ternary complex components are critical for dynein-dependent spindle positioning in HeLa cells and C. elegans embryos. Moreover, using mutant versions of Gα in both systems, we established that dynein acts at the membrane to direct spindle positioning. Importantly, we identified a region within NuMA that mediates association with dynein. By using this region to target dynein to the plasma membrane, we demonstrated that the mere presence of dynein at that location is sufficient to direct spindle positioning in HeLa cells. Overall, we propose a model in which the ternary complex serves to anchor dynein at the plasma membrane to ensure correct spindle positioning.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-α-tubulina monoclonal antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Dynein (Intermediate Chain) antibody produced in mouse, clone 70.1, ascites fluid