Merck
SLC25A40 Facilitates Anticancer Drug Resistance in Human Leukemia K562 Cells.
Biological & pharmaceutical bulletin (2023-07-06)
Nodoka Kudo, Rikuma Kouno, Yoshihiko Shibayama
RESUMEN
The chronic myelogenous leukemia cell line, K562/ADM is derived from the K562 cell line, which is resistant to doxorubicin (alias, adriamycin: ADM). P-glycoprotein levels are significantly higher in K562/ADM cells than in K562 cells. The overexpression of p-glycoprotein has been shown to cause drug resistance. Therefore, the present study investigated a novel mechanism underlying the drug resistance of K562/ADM cells. A gene ontology analysis demonstrated that the expression of solute carrier (SLC)-mediated transmembrane transport genes was significantly higher in K562/ADM cells than in K562 cells. The expression level of a member of the SLC family, SLC25A40 was higher in K562/ADM cells than in K562 cells. SLC25A40 is located near the ABCB1 gene. A real-time PCR analysis showed that the expression of SLC25A40, ABCB4, and ADAM22 was up-regulated. These genes are located close to SLC25A40. The down-regulation of SLC25A40 significantly decreased the mitochondrial concentration of glutathione and cell proliferation. Collectively, the present results demonstrated that the expression of SLC25A40 was up-regulated in K562/ADM cells, which enhanced to cell proliferation, and that the expression of SLC25A40 affected drug resistance to ADM.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-SLC25A40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution