Merck
The sodium channel subunit SCNN1B suppresses colorectal cancer via suppression of active c-Raf and MAPK signaling cascade.
Oncogene (2022-12-24)
Yun Qian, Lianxin Zhou, Simson Tsz Yat Luk, Jiaying Xu, Weilin Li, Hongyan Gou, Huarong Chen, Wei Kang, Jun Yu, Chi Chun Wong
RESUMEN
The incidence of colorectal cancer (CRC) is rising worldwide. Here, we identified SCNN1B as an outlier down-regulated in CRC and it functions as a tumor suppressor. SCNN1B mRNA and protein expression were down-regulated in primary CRC and CRC cells. In a tissue microarray cohort (N = 153), SCNN1B protein was an independent prognostic factor for favorable outcomes in CRC. Ectopic expression of SCNN1B in CRC cell lines suppressed cell proliferation, induced apoptosis, and cell cycle arrest, and suppressed cell migration in vitro. Xenograft models validated tumor suppressive function of SCNN1B in vivo. Mechanistically, Gene Set Enrichment Analysis (GSEA) showed that SCNN1B correlates with KRAS signaling. Consistently, MAPK qPCR and kinase arrays revealed that SCNN1B suppressed MAPK signaling. In particular, SCNN1B overexpression suppressed p-MEK/p-ERK expression and SRE-mediated transcription activities, confirming blockade of Ras-Raf-MEK-ERK cascade. Mechanistically, SCNN1B did not affect KRAS activation, instead impairing activation of c-Raf by inducing its inhibitory phosphorylation and targeting active c-Raf for degradation. The ectopic expression of c-Raf fully rescued cell proliferation and colony formation in SCNN1B-overexpressing CRC cells, confirming c-Raf as the principal molecular target of SCNN1B. In summary, we identified SCNN1B as a tumor suppressor by functioning as a c-Raf antagonist, which in turn suppressed oncogenic MEK-ERK signaling.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-SCNN1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution