Merck
Molecular basis of MHC I quality control in the peptide loading complex.
Nature communications (2022-08-11)
Alexander Domnick, Christian Winter, Lukas Sušac, Leon Hennecke, Mario Hensen, Nicole Zitzmann, Simon Trowitzsch, Christoph Thomas, Robert Tampé
RESUMEN
Major histocompatibility complex class I (MHC I) molecules are central to adaptive immunity. Their assembly, epitope selection, and antigen presentation are controlled by the MHC I glycan through a sophisticated network of chaperones and modifying enzymes. However, the mechanistic integration of the corresponding processes remains poorly understood. Here, we determine the multi-chaperone-client interaction network of the peptide loading complex (PLC) and report the PLC editing module structure by cryogenic electron microscopy at 3.7 Å resolution. Combined with epitope-proofreading studies of the PLC in near-native lipid environment, these data show that peptide-receptive MHC I molecules are stabilized by multivalent chaperone interactions including the calreticulin-engulfed mono-glucosylated MHC I glycan, which only becomes accessible for processing by α-glucosidase II upon loading of optimal epitopes. Our work reveals allosteric coupling between peptide-MHC I assembly and glycan processing. This inter-process communication defines the onset of an adaptive immune response and provides a prototypical example of the tightly coordinated events in endoplasmic reticulum quality control.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-polihistidina monoclonal antibody produced in mouse, clone HIS-1, ascites fluid
Sigma-Aldrich
Anti-Calreticulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution