Merck
A perspective on the use of iTRAQ reagent technology for protein complex and profiling studies.
Journal of experimental botany (2006-04-01)
Lynn R Zieske
RESUMEN
Proteomic research includes the characterization of protein mixtures in order to understand complex biological systems and determine relationships between proteins, their function, and protein-protein interactions. Often the goal of such research is to monitor changes of proteins in perturbed systems, a type of study referred to as differential expression analysis. To perform these studies requires the ability to execute some type of differential comparison of a given protein state in reference to some type of a control. The iTRAQ reagents are a set of isobaric reagents which are amine specific and allow for the identification and quantitation of up to four different samples simultaneously. The amine specificity of these reagents makes most peptides in a sample amenable to this labeling strategy with no loss of information from samples involving post-translational modifications, such as the scrutiny of signal transduction pathways that often involve phosphorylation phenomena. In addition, the multiplexing capacity of these reagents allows for information replication within certain LC-MS/MS experimental regimes, providing additional statistical validation within any given experiment. The results presented herein demonstrate a few examples of the wide variety of quantitative information that can be realized when undertaking such experimental approaches. These include temporal analysis of drug-induced-protein expression, discovery and elucidation of disease markers, and protein-protein interactions in multi-protein complexes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
iTRAQ® Reagent - Multiplex Buffer Kit