Merck
Application of RNA interference in tick salivary gland research.
Journal of biomolecular techniques : JBT (2006-03-09)
Vijay G Ramakrishnan, Majd N Aljamali, John R Sauer, Richard C Essenberg
RESUMEN
Ticks are obligate ectoparasites that feed on a variety of hosts including mammals, birds and reptiles. Prolonged attachment on the host and an ability to transmit a wide variety of pathogens are the special features of tick feeding. Salivary glands are the major route for secretion of excess fluid, several proteins, and factors that counteract the host immune response and hence play a significant role in the success of tick feeding. RNA interference (RNAi) enables scientists to silence genes encoding proteins in an absolutely sequence specific manner at the mRNA level. This technique has already been successfully employed in analyzing roles of proteins of important functions or in assigning roles to several proteins of unknown functions in a variety of animals. In this review, we outline the process of RNAi and the applicability of RNAi in tick salivary gland research.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
α-Naphthyl Butyrate Solution