Merck
  • Experimental Helicobacter pylori infection induces antral gastritis and gastric mucosa-associated lymphoid tissue in guinea pigs.
Experimental Helicobacter pylori infection induces antral gastritis and gastric mucosa-associated lymphoid tissue in guinea pigs.
Infection and immunity (1998-05-29)
N H Shomer, C A Dangler, M T Whary, J G Fox
RESUMEN
Humans infected with Helicobacter pylori have abnormally low levels of the antioxidant vitamin C, which protects against the formation of carcinogenic nitrosamines, in gastric juice. Guinea pigs, like humans and nonhuman primates, have a dietary requirement for vitamin C. As such, these species have gastrointestinal vitamin C transport systems not found in other animals. We have developed and characterized a guinea pig model of chronic gastric H. pylori infection with the rodent-adapted Sydney strain of H. pylori. At 4 weeks postinfection, five of six animals of the infected group and zero of two animals of the control group were positive for H. pylori as determined by culture or PCR. At 15 weeks, six of six animals of the infected group and zero of two animals of the control group were positive. H. pylori-specific seroconversion was observed among infected animals. There were no histologic abnormalities in the gastric antra or fundi of control guinea pigs. In contrast, there was multifocal, mild to moderate lymphohistiocytic antral gastritis and formation of antral lymphoid follicles in H. pylori-infected animals. The lesion distribution in the gastric antra paralleled that observed in H. pylori-infected humans. The H. pylori-infected guinea pig should prove useful in modeling the interaction of helicobacter and vitamin C in gastric carcinogenesis.