Merck
Conformational rearrangement during activation of a metabotropic glutamate receptor.
Nature chemical biology (2021-01-06)
Brandon Wey-Hung Liauw, Hamid Samareh Afsari, Reza Vafabakhsh
RESUMEN
G protein-coupled receptors (GPCRs) relay information across cell membranes through conformational coupling between the ligand-binding domain and cytoplasmic signaling domain. In dimeric class C GPCRs, the mechanism of this process, which involves propagation of local ligand-induced conformational changes over 12 nm through three distinct structural domains, is unknown. Here, we used single-molecule FRET and live-cell imaging and found that metabotropic glutamate receptor 2 (mGluR2) interconverts between four conformational states, two of which were previously unknown, and activation proceeds through the conformational selection mechanism. Furthermore, the conformation of the ligand-binding domains and downstream domains are weakly coupled. We show that the intermediate states act as conformational checkpoints for activation and control allosteric modulation of signaling. Our results demonstrate a mechanism for activation of mGluRs where ligand binding controls the proximity of signaling domains, analogous to some receptor kinases. This design principle may be generalizable to other biological allosteric sensors.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder