Merck
THZ1 Reveals Roles for Cdk7 in Co-transcriptional Capping and Pausing.
Molecular cell (2015-08-11)
Kyle A Nilson, Jiannan Guo, Michael E Turek, John E Brogie, Elizabeth Delaney, Donal S Luse, David H Price
RESUMEN
The Cdk7 subunit of TFIIH phosphorylates RNA polymerase II (Pol II) during initiation, and, while recent studies show that inhibition of human Cdk7 negatively influences transcription, the mechanisms involved are unclear. Using in vitro transcription with nuclear extract, we demonstrate that THZ1, a covalent Cdk7 inhibitor, causes defects in Pol II phosphorylation, co-transcriptional capping, promoter proximal pausing, and productive elongation. THZ1 does not affect initiation but blocks essentially all Pol II large subunit C-terminal domain (CTD) phosphorylation. We found that guanylylation of nascent RNAs is length dependent and modulated by a THZ1-sensitive factor present in nuclear extract. THZ1 impacts pausing through a capping-independent block of DSIF and NELF loading. The P-TEFb-dependent transition into productive elongation was also inhibited by THZ1, likely due to loss of DSIF. Capping and pausing were also reduced in THZ1-treated cells. Our results provide mechanistic insights into THZ1 action and how Cdk7 broadly influences transcription and capping.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-2,2,7-Trimethylguanosine Mouse mAb (K121) Agarose Conjugate, suspension (Liquid), clone K121, Calbiochem®