Merck
  • Comparative Analyses of Four Chemicals Used to Control Black Mold Disease in Tomato and Its Effects on Defense Signaling Pathways, Productivity and Quality Traits.
Comparative Analyses of Four Chemicals Used to Control Black Mold Disease in Tomato and Its Effects on Defense Signaling Pathways, Productivity and Quality Traits.
Plants (Basel, Switzerland) (2020-07-02)
Hoda A S El-Garhy, Fayz A Abdel-Rahman, Abdelhakeem S Shams, Gamal H Osman, Mahmoud M A Moustafa
RESUMEN
The field application of safe chemical inducers plays a vital role in the stimulation of systematic acquired resistance (SAR) of plants. In this study, the efficacy use of three and six field applications with chitosan, lithovit, and K-thiosulfate at 4 gL-1 and salicylic acid at 1.5 gL-1 in improving tomato productivity, quality, and modifying the defense signaling pathways to the Alternaria alternata infection was investigated. Salicylic acid was the most effective in vitro where it completely inhibited the growth of Alternaria alternata. The highest yield quantity was recorded with six applications with Chitosan followed by Salicylic acid; also, they were the most effective treatments in controlling the Alternaria alternata infection in tomato fruits. The maximum increase in chitinase and catalase activity of tomato fruits was observed at five days after inoculation, following treatment with six sprays of salicylic acid followed by chitosan. The transcript levels of seven defense-related genes: ethylene-responsive transcription factor 3 (RAP), xyloglucan endotransglucosylase 2 (XET-2), catalytic hydrolase -2 (ACS-2), proteinase inhibitor II (PINII), phenylalanine ammonia-lyase 5 (PAL5), lipoxygenase D (LOXD), and pathogenesis-related protein 1 (PR1) were upregulated in response to all treatments. The highest expression levels of the seven studied genes were recorded in response to six foliar applications with chitosan. Chitosan followed by salicylic acid was the most effective among the tested elicitors in controlling the black mold rot in tomato fruits. In conclusion, pre-harvest chitosan and salicylic acid in vivo application with six sprays could be recommended as effective safe alternatives to fungicides against black mold disease in tomato fruits.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Potassium thiosulfate, ≥95%