Merck
Itch inhibits IL-17-mediated colon inflammation and tumorigenesis by ROR-γt ubiquitination.
Nature immunology (2016-06-21)
Mahesh Kathania, Prashant Khare, Minghui Zeng, Brandi Cantarel, Haiying Zhang, Hideki Ueno, K Venuprasad
RESUMEN
Dysregulated expression of interleukin 17 (IL-17) in the colonic mucosa is associated with colonic inflammation and cancer. However, the cell-intrinsic molecular mechanisms by which IL-17 expression is regulated remain unclear. We found that deficiency in the ubiquitin ligase Itch led to spontaneous colitis and increased susceptibility to colon cancer. Itch deficiency in the TH17 subset of helper T cells, innate lymphoid cells and γδ T cells resulted in the production of elevated amounts of IL-17 in the colonic mucosa. Mechanistically, Itch bound to the transcription factor ROR-γt and targeted ROR-γt for ubiquitination. Inhibition or genetic inactivation of ROR-γt attenuated IL-17 expression and reduced spontaneous colonic inflammation in Itch(-/-) mice. Thus, we have identified a previously unknown role for Itch in regulating IL-17-mediated colonic inflammation and carcinogenesis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
SR1001, ≥98% (HPLC)