Merck
Inhibition of hepatitis C virus replication in vitro by xanthohumol, a natural product present in hops.
Planta medica (2013-12-21)
Sai Lou, Yi-Min Zheng, Shan-Lu Liu, Jianming Qiu, Qunying Han, Na Li, Qianqian Zhu, Pingping Zhang, Cuiling Yang, Zhengwen Liu
RESUMEN
Hepatitis C virus is a major cause of chronic liver disease worldwide. Xanthohumol, a prenylated flavonoid from hops, has various biological activities including an antiviral effect. It was previously characterized as a compound that inhibits bovine viral diarrhea virus, a surrogate model of hepatitis C virus. In the present work, xanthohumol was examined for its ability to inhibit hepatitis C virus replication in a cell culture system carrying replicating hepatitis C virus RNA replicon. 0.2 % DMSO and 500 units/mL interferon-alpha treatments were set as a negative and positive control, respectively. The inhibitory effect by xanthohumol was determined by the luciferase activity of the infected Huh7.5 cell lysates and the hepatitis C virus RNA levels in the culture. Xanthohumol at 3.53 µM significantly decreased the luciferase activity compared to the negative control (p < 0.01). Xanthohumol at 7.05 µM further decreased the luciferase activity compared to xanthohumol at 3.53 µM (p = 0.015). Xanthohumol at 7.05 µM or 14.11 µM achieved an inhibitory effect similar to that of interferon-alpha 2b (p > 0.05). Xanthohumol at 3.53 µM significantly reduced the hepatitis C virus RNA level compared to the negative control (p = 0.001). Although the results of xanthohumol at 7.05 µM had a higher variation, xanthohumol at the 7.05 µM and 14.11 µM decreased the hepatitis C virus RNA level to that achieved by interferon-alpha (p > 0.05). In conclusion, xanthohumol displays anti-hepatitis C virus activity in a cell culture system and may be potentially used as an alternative or complementary treatment against the hepatitis C virus.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Xanthohumol from hop (Humulus lupulus), ≥96% (HPLC)