Merck
2-(4-Ethoxy phenyl)-4-phenyl quinoline organic phosphor for solution processed blue organic light-emitting diodes.
Luminescence : the journal of biological and chemical luminescence (2018-06-01)
Minakshi Ghate, N Thejo Kalyani, S J Dhoble
RESUMEN
This paper reports the synthesis and characterization of 2-(4-ethoxyphenyl)-4-phenyl quinoline (OEt-DPQ) organic phosphor using an acid-catalyzed Friedlander reaction and the preparation of blended thin films by molecularly doping OEt-DPQ in poly(methyl methacrylate) (PMMA) at different wt%. The molecular structure of the synthesized phosphor was confirmed by Fourier transform infra-red (FTIR) spectroscopy and nuclear magnetic resonance spectra (NMR). Surface morphology and percent composition of the elements were assessed by scanning electron microscopy (SEM) and energy dispersive analysis of X-rays (EDAX). The thermal stability and melting point of OEt-DPQ and thin films were probed by thermo-gravimetric analysis (TGA)/differential thermal analysis (DTA) and were found to be 80°C and 113.6°C, respectively. UV-visible optical absorption spectra of OEt-DPQ in the solid state and blended films produced absorption bands in the range 260-340 nm, while photoluminescence (PL) spectra of OEt-DPQ in the solid state and blended thin films demonstrated blue emission that was registered at 432 nm when excited at 363-369 nm. However, solvated OEt-DPQ in chloroform, tetrahydrofuran or dichloromethane showed a blue shift of 31-43 nm. Optical absorption and emission parameters such as molar extinction coefficient (ε), energy gap (Eg ), transmittance (T), reflectance (R), refractive index (n), oscillator energy (E0 ) and oscillator strength (f), quantum yield (φf ), oscillator energy (E0 ), dispersion energy (Ed ), Commission Internationale de l'Éclairage (CIE) co-ordinates and energy yield fluorescence (EF ) were calculated to assess the phosphor's suitability as a blue emissive material for opto-electronic applications such as organic light-emitting diodes (OLEDs), flexible displays and solid-state lighting technology.