Merck
  • Network pharmacology study reveals energy metabolism and apoptosis pathways-mediated cardioprotective effects of Shenqi Fuzheng.
Network pharmacology study reveals energy metabolism and apoptosis pathways-mediated cardioprotective effects of Shenqi Fuzheng.
Journal of ethnopharmacology (2018-08-27)
Jie Liao, Cui Hao, Wenhua Huang, Xin Shao, Yangang Song, Liangfeng Liu, Ni Ai, Xiaohui Fan
RESUMEN
Shenqi Fuzheng (SQ) is a renowned traditional Chinese medicine extracted from Radix Codonopsis and Radix Astragali. Although SQ is widely used to treat myocardial ischemia-reperfusion (I/R) injury, the molecular mechanisms supporting its clinical application remain elusive. The purpose of current study was to understand its cardioprotective effects at the molecular level using network pharmacology approach. In an I/R injury animal model, the beneficial pharmacological activities of SQ were confirmed by decreased infarct range observed on drug treated rats versus control group. Additionally, several serum biochemical indicators were in concord with this observation. Subsequently, a microarray experiment was performed to reveal the influence on injured heart at the gene expression level by this TCM injection. We then proposed a network analysis algorithm NTRA to discover the key nodes based on both disease network structure and transcriptomics. Using NRIODN, a method developed by our group previously, the holistic changes on the gene network induced by for I/R injury and SQ treatment were evaluated. Pathway enrichment analysis of highly ranked genes by NTRA showed that PPAR and apoptosis pathways were highly related to I/R injury. Finally, western blot results showed increased level of the PPARα and BAX protein in the heart after injection treatment which confirmed the hypothesis. In conclusion, our results suggest that SQ injection exerts protective effect against myocardial ischemia-reperfusion injury through multiple pathways, including myocardial energy metabolism improvement, cell adhesion inhibition, inflammatory reaction perturbation, myocardial apoptosis reduction and ventricular remodeling avoidance.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Human MB / Myoglobin ELISA Kit