Merck
Progesterone receptor membrane component 1 modulates human cytochrome p450 activities in an isoform-dependent manner.
Drug metabolism and disposition: the biological fate of chemicals (2011-08-10)
Shingo Oda, Miki Nakajima, Yasuyuki Toyoda, Tatsuki Fukami, Tsuyoshi Yokoi
RESUMEN
Cytochromes P450 (P450s) catalyze the metabolism of a wide spectrum of compounds. Recently, progesterone receptor membrane component 1 (PGRMC1), which shares a key structural motif with cytochrome b(5), has been reported to bind to sterol- or steroid-synthesizing P450s, enhancing their activities. In this study, we investigated whether PGRMC1 affects human drug-metabolizing P450 activities. Using coexpression systems for PGRMC1 and P450s (CYP3A4, CYP2C9, or CYP2E1) in HepG2 cells, we found that PGRMC1 decreased the V(max) values and increased the K(m) values of the CYP3A4 activities, and it decreased the V(max) values but did not affect the K(m) values of the CYP2C9 activities. In contrast, PGRMC1 hardly affected the CYP2E1 activities. These results suggest that PGRMC1 negatively modulates the drug-metabolizing activities of P450, although it was isoform but not substrate dependent. It is worth noting that coimmunoprecipitation analysis using coexpression systems for FLAG-PGRMC1 and Myc-P450s in human embryonic kidney 293 cells revealed that PGRMC1 interacts with all three P450s, although the affinity seemed to vary. In 29 human liver microsomes (HLMs), there was a 5-fold variability in the PGRMC1 protein levels. By the correlation analyses using the P450 activities and the PGRMC1 levels, we could neither observe the contribution of PGRMC1 to the P450 activities in HLMs nor that of the NADPH-cytochrome P450 reductase or cytochrome b(5). In conclusion, in contrast to sterol- or steroid-synthesizing P450s, we found that PGRMC1 negatively modulates the human drug-metabolizing activities of P450 through direct interaction. Further studies are needed to determine the clinical significance of PGRMC1 in the pharmacokinetics of drugs.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
ANTI-FLAG® M2 monoclonal antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-PGRMC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution