Merck
  • The cytotoxic effect of Bowman-Birk isoinhibitors, IBB1 and IBBD2, from soybean (Glycine max) on HT29 human colorectal cancer cells is related to their intrinsic ability to inhibit serine proteases.
The cytotoxic effect of Bowman-Birk isoinhibitors, IBB1 and IBBD2, from soybean (Glycine max) on HT29 human colorectal cancer cells is related to their intrinsic ability to inhibit serine proteases.
Molecular nutrition & food research (2009-11-04)
Alfonso Clemente, Francisco Javier Moreno, Maria del Carmen Marín-Manzano, Elisabeth Jiménez, Claire Domoney
ABSTRACT
Bowman-Birk inhibitors (BBI) from soybean and related proteins are naturally occurring protease inhibitors with potential health-promoting properties within the gastrointestinal tract. In this work, we have investigated the effects of soybean BBI proteins on HT29 colon adenocarcinoma cells, compared with non-malignant colonic fibroblast CCD-18Co cells. Two major soybean isoinhibitors, IBB1 and IBBD2, showing considerable amino acid sequence divergence within their inhibitory domains, were purified in order to examine their functional properties, including their individual effects on the proliferation of HT29 colon cancer cells. IBB1 inhibited both trypsin and chymotrypsin whereas IBBD2 inhibited trypsin only. Despite showing significant differences in their enzyme inhibitory properties, the median inhibitory concentration values determined for IBB1 and IBBD2 on HT29 cell growth were not significantly different (39.9+/-2.3 and 48.3+/-3.5 microM, respectively). The cell cycle distribution pattern of HT29 colon cancer cells was affected by BBI treatment in a dose-dependent manner, with cells becoming blocked in the G0-G1 phase. Chemically inactive soybean BBI had a weak but non-significant effect on the proliferation of HT29 cells. The anti-proliferative properties of BBI isoinhibitors from soybean reveal that both trypsin- and chymotrypsin-like proteases involved in carcinogenesis should be considered as potential targets of BBI-like proteins.
Sigma-Aldrich
Trypsin-chymotrypsin inhibitor from Glycine max (soybean), lyophilized powder