- Rotational diffusion of magnetic nanoparticles in protein solutions.
Rotational diffusion of magnetic nanoparticles in protein solutions.
Journal of colloid and interface science (2017-07-28)
Ana C Bohórquez, Chuncheng Yang, Donald Bejleri, Carlos Rinaldi
PMID28750242
ABSTRACT
The rotational diffusion of polyethylene glycol coated magnetic nanoparticles in serum albumin solutions was investigated in a range spanning 0mgmL
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, powder, suitable for cell culture