Merck
  • Characterization of vector-based delivery of neurogenin-3 in murine diabetes.
Characterization of vector-based delivery of neurogenin-3 in murine diabetes.
Human gene therapy (2014-03-19)
Neil Phillips, Mark A Kay
ABSTRACT
Treatment of type 1 diabetes with gene transfer-induced cellular reprogramming requires a pancreatic transcription factor such as Neurogenin-3 (Ngn3) and as of yet unknown component of the adenoviral particle. Despite intensive study, there are many unsolved processes related to the mechanisms and physiological parameters related to diabetes correction using this approach. While we confirm that systemic delivery of adenovirus (Ad)-Ngn3 provides long-lasting correction of streptozotocin (STZ)-induced hyperglycemia and restoration of growth curves, we found that insulin levels and glucose tolerance tests are not fully restored. By altering the innate and antigen-specific immune responses, we establish that the former likely plays some role in the reprogramming process. Interestingly, Ad-hNgn3 therapy in diabetic animals appeared to protect them from secondary STZ challenge. The resistance to secondary STZ response was more pronounced at later time points, indicating that a period of cell maturation and/or expansion may be required in order to promote lasting correction. More importantly, these results suggest that the long-term reprogrammed cells are not fully reprogrammed into β-cells, which in the case of autoimmune diabetes may be advantageous in a long-term treatment strategy. Finally, we show that the prophylactic administration of Ad-hNgn3 before diabetic induction protected mice from developing hyperglycemia, demonstrating the potential for reducing or eliminating disease progression should treatment be initiated early or before onset of symptoms.
Sigma-Aldrich
Dimethyldioctadecylammonium bromide, ≥98% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Gadolinium(III) chloride hexahydrate, 99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyinosinic–polycytidylic acid sodium salt, TLR ligand tested