Merck
  • Proliferative activity of rectal mucosa and soluble fecal bile acids in patients with normal colons and in patients with colonic polyps or cancer.
Proliferative activity of rectal mucosa and soluble fecal bile acids in patients with normal colons and in patients with colonic polyps or cancer.
Cancer letters (1988-01-01)
J Stadler, K S Yeung, R Furrer, N Marcon, H S Himal, W R Bruce
ABSTRACT
Rectal biopsies and fecal collections were obtained from a consecutive series of 34 outpatients prior to colonoscopy at a gastroenterology clinic. Subsequently, 14 were found to have no colonic pathology, 13 had adenomatous polyps, (3 of those had a previous history of colon cancer) and 7 were diagnosed with colon cancer. In confirmation of earlier studies the tritiated thymidine labelling index was higher in patients with tumors than in those without pathology (7.9% vs. 5.8% with P = 0.06). The patients with colonic tumors also had significantly higher levels of deoxycholic acid (P = 0.01) and lithocholic acid (P = 0.005) in the aqueous extract of their feces. This study shows that these biochemical measures may indicate colon cancer risk.
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)