Merck
  • Activation of morphine glucuronidation by fatty acyl-CoAs and its plasticity: a comparative study in humans and rodents including chimeric mice carrying human liver.
Activation of morphine glucuronidation by fatty acyl-CoAs and its plasticity: a comparative study in humans and rodents including chimeric mice carrying human liver.
Drug metabolism and pharmacokinetics (2010-07-09)
Arief Nurrochmad, Yuji Ishii, Hitomi Nakanoh, Tae Inoue, Toru Horie, Kazumi Sugihara, Shigeru Ohta, Akinobu Taketomi, Yoshihiko Maehara, Hideyuki Yamada
ABSTRACT
The formation of morphine-3-glucuronide (M-3-G, pharmacologically inactive) and morphine-6-glucuronide (M-6-G, active metabolite) by liver microsomes from humans and rodents, including chimeric mice carrying human liver, was evaluated in the presence of fatty acyl-CoAs. Medium- to long-chain fatty acyl-CoAs, including oleoyl-CoAs, at a physiologic level (around 15 microM) markedly enhanced M-3-G formation catalyzed by rat liver microsomes. A separate experiment indicated that 15 microM oleoyl-CoA enhanced (14)C-UDP-glucuronic acid (UDPGA) uptake by microsomes. The activation by acyl-CoAs disappeared or was greatly reduced by either pre-treating microsomes with detergent or freezing/thawing the rat liver before preparation. Many of the microsomes prepared from frozen human livers (N=14) resisted oleoyl-CoA-mediated activation of UDP-glucuronosyltransferase (UGT) activity, including M-6-G formation, which is highly specific to humans. In sharp contrast, the activity of M-6-G and M-3-G formation in freshly-prepared hepatic microsomes from chimeric mice with humanized liver was potently activated by oleoyl-CoA. Thus, acyl-CoAs activate morphine glucuronidation mediated by human as well as rat UGTs. This activation is assumed to be due to the acyl-CoA-facilitated transportation of UDPGA, and microsomes need to maintain the intact conditions required for the activation. The function of UGT appears to be dynamically changed depending on the cellular acyl-CoA level in many species.
Product Description
Supelco
Morphine-3-β-D-glucuronide solution, 1 mg/mL in methanol with 0.05% NaOH, ampule of 1 mL, certified reference material, Cerilliant®
SKU
Pack Size
Availability
Price
Quantity
Supelco
Morphine-3-β-D-glucuronide solution, 100 μg/mL in methanol: water ( 50:50(v/v)), ampule of 1 mL, certified reference material, Cerilliant®