Merck
  • Rapid simultaneous determination of 14 sulfonamides in wastewater by liquid chromatography tandem mass spectrometry.
Rapid simultaneous determination of 14 sulfonamides in wastewater by liquid chromatography tandem mass spectrometry.
Journal of separation science (2007-09-01)
Sai Ye, Ziwei Yao, Guangshui Na, Juying Wang, Deyi Ma
ABSTRACT
This paper describes a rapid method for the determination of 14 kinds of sulfonamides (SAs) in wastewater using SPE, and LC-MS/MS with positive ESI (ESI(+)) and selected reaction monitoring (SRM) mode. The SPE was performed on an Oasis hydrophilic-lipophilic-balanced (HLB) cartridge. Chromatographic separation on a C18 column was achieved using a binary eluent containing methanol and water with 0.2% formic acid. Typical recoveries of the analytes ranged from 22.3 to 87.0% at a fortification level of 100 ng/L. The LODs in wastewater except sulfathiazole (3 ng/L) could be detected and quantified at levels as low as 1 ng/L. Finally, the method was applied to water from the municipal outlet and the aquaculture wastewater effluent. Sulfamethazine (SM(2)), sulfamethoxypyridazine (SMP), and sulfamethoxazole (SMZ) were most frequently found in wastewater in a concentration range between 1.2 and 31.7 ng/L.
Product Description
Supelco
Sulfamethoxypyridazine, VETRANAL®, analytical standard
Supelco
Sulfamethoxypyridazine, analytical standard