Merck
  • Structure of the eukaryotic thiamine pyrophosphate riboswitch with its regulatory ligand.
Structure of the eukaryotic thiamine pyrophosphate riboswitch with its regulatory ligand.
Science (New York, N.Y.) (2006-05-06)
Stéphane Thore, Marc Leibundgut, Nenad Ban
ABSTRACT
Riboswitches are untranslated regions of messenger RNA, which adopt alternate structures depending on the binding of specific metabolites. Such conformational switching regulates the expression of proteins involved in the biosynthesis of riboswitch substrates. Here, we present the 2.9 angstrom-resolution crystal structure of the eukaryotic Arabidopsis thaliana thiamine pyrophosphate (TPP)-specific riboswitch in complex with its natural ligand. The riboswitch specifically recognizes the TPP via conserved residues located within two highly distorted parallel "sensor" helices. The structure provides the basis for understanding the reorganization of the riboswitch fold upon TPP binding and explains the mechanism of resistance to the antibiotic pyrithiamine.
Sigma-Aldrich
Pyrithiamine hydrobromide, ~95%, crystalline
Pricing and availability is not currently available.