Merck
  • Deciphering the Role of Leptospira Surface Protein LigA in Modulating the Host Innate Immune Response.
Deciphering the Role of Leptospira Surface Protein LigA in Modulating the Host Innate Immune Response.
Frontiers in immunology (2022-01-04)
Ajay Kumar, Vivek P Varma, Kavela Sridhar, Mohd Abdullah, Pallavi Vyas, Muhammed Ashiq Thalappil, Yung-Fu Chang, Syed M Faisal
ABSTRACT
Leptospira, a zoonotic pathogen, is known to infect various hosts and can establish persistent infection. This remarkable ability of bacteria is attributed to its potential to modulate (activate or evade) the host immune response by exploiting its surface proteins. We have identified and characterized the domain of the variable region of Leptospira immunoglobulin-like protein A (LAV) involved in immune modulation. The 11th domain (A11) of the variable region of LigA (LAV) induces a strong TLR4 dependent innate response leading to subsequent induction of humoral and cellular immune responses in mice. A11 is also involved in acquiring complement regulator FH and binds to host protease Plasminogen (PLG), there by mediating functional activity to escape from complement-mediated killing. The deletion of A11 domain significantly impaired TLR4 signaling and subsequent reduction in the innate and adaptive immune response. It also inhibited the binding of FH and PLG thereby mediating killing of bacteria. Our study discovered an unprecedented role of LAV as a nuclease capable of degrading Neutrophil Extracellular Traps (NETs). This nuclease activity was primarily mediated by A11. These results highlighted the moonlighting function of LigA and demonstrated that a single domain of a surface protein is involved in modulating the host innate immune defenses, which might allow the persistence of Leptospira in different hosts for a long term without clearance.
Sigma-Aldrich
Anti-Complement Factor H antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
UPA human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)