
Anhydrous Sodium Succinate

Synonym(s):
Sodium succinate dibasic, Anhydrous butanedioic acid disodium salt, Succinic acid disodium salt
Linear Formula:
NaOOCCH2CH2COONa
CAS Number:
Molecular Weight:
162.05
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2