Skip to Content
Merck
2,6-Dichloro-1-fluoropyridinium tetrafluoroborate 97%

2,6-Dichloro-1-fluoropyridinium tetrafluoroborate

2,6-Dichloro-1-fluoropyridinium tetrafluoroborate 97%
Empirical Formula (Hill Notation):
C5H3BCl2F5N
CAS Number:
Molecular Weight:
253.79
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing