1535788
USP
Phosphatidylinositol (Soy) Sodium
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Phosphatidylinositols, soya, sodium salts
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
phosphatidylinositol
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
−20°C
SMILES string
[Na+].[P](=O)([O-])(O[C@@H]1C([C@@H](C(C(C1O)O)O)O)O)OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC
InChI
1S/C43H79O13P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(45)55-35(34-54-57(51,52)56-43-41(49)39(47)38(46)40(48)42(43)50)33-53-36(44)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,35,38-43,46-50H,3-10,12,14-16,19-34H2,1-2H3,(H,51,52);/q;+1/p-1/b13-11-,18-17-;/t35-,38?,39-,40?,41?,42?,43-;/m1./s1
InChI key
DCDQYHYLAGWAPO-UQZYTPKFSA-M
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also used to prepare Phospholipids standard stock solution during the assay of Lecithin by using the liquid chromatography method coupled with an evaporative light-scattering detector.
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service