PHL82602
7α,β-Morroniside
phyproof® Reference Substance
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
format
neat
storage temp.
−20°C
SMILES string
O=C(C1=CO[C@H]([C@]2([C@@]1(CC(O[C@H]2C)O)[H])[H])O[C@@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO)OC
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG.
This reference substance is the naturally occurring mixture of 7alpha-morroniside (CAS # 61849-88-5) and 7beta-morroniside (CAS # 25406-64-8). At this moment there is no CAS # available for the diastereomeric mixture. The two diastereomers cannot be separated chromatographically on regular columns. The 13C-NMR data, however, confirms the presence of both diastereomes in similar ratios.
This reference substance is the naturally occurring mixture of 7alpha-morroniside (CAS # 61849-88-5) and 7beta-morroniside (CAS # 25406-64-8). At this moment there is no CAS # available for the diastereomeric mixture. The two diastereomers cannot be separated chromatographically on regular columns. The 13C-NMR data, however, confirms the presence of both diastereomes in similar ratios.
Other Notes
This compound is commonly found in plants of the genus: sambucus
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service