Skip to Content
Merck
All Photos(1)

Key Documents

36082

Supelco

TraceCERT® Multiresidue Pesticide Standard Mix #4

in acetonitrile, TraceCERT®, (Manufactured by: Sigma-Aldrich Production GmbH, Switzerland)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

product line

TraceCERT®

grade

TraceCERT®

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

(Manufactured by: Sigma-Aldrich Production GmbH, Switzerland)

concentration

in acetonitrile
100 μg/mL each component in acetonitrile

application(s)

cannabis testing
food and beverages
(pesticide residue testing)

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Other Notes

8 components. When combined with other mixes, suitable for use in meeting California state requirements for pesticide testing of cannabis products
Check out our entire range of pesticide mixes for Cannabis testing

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Oxamyl
    Carbaryl
    Methomyl
    Dimethomorph
    Metalaxyl
    Imidacloprid
    Thiamethoxam
    Chlorantraniliprole
    See All (8)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service