Skip to Content
Merck
All Photos(1)

Key Documents

22562

Supelco

Methane (99.0%)

99.0%, cylinder of 14 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12000000
Pricing and availability is not currently available.

grade

analytical standard

Assay

99.0%

form

gas

packaging

cylinder of 14 L

manufacturer/tradename

Scott Gas T109-14

availability

not available in Canada, EU

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

Related Categories

General description

Specifications:
Contents
: 14 liters
Pressure: 240 psig
Outlet fitting: CGA-160-1/8 inch, NPT F
Weight: 1.5 lb.
Dimensions
: 3 x 11 inches
D.O.T. specifications: 4B240

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

comparable product

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-306.4 °F - closed cup

Flash Point(C)

-188 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

https://pubchem.ncbi.nlm.nih.gov/compound/methane#section=GC-MS
National Institutes of Health Bulletin null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service