Key Documents
Z757497
Sartorius Proline® Plus multichannel mechanical pippettor
12-channel, volume 30-300 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
feature
12-channel
electronic: no
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 728240
volume
30-300 μL
General description
The Proline® Plus 12 channel mechanical pipettes offer comfort and quality for everyday manual pipetting. It incorporates durable construction with ease and lightness of use. The multichannel pipette consists of Optiload spring-loaded tip cones that are ideal for loading and ejecting the tips. It also seals the tips on all channels equally with minimal force.
Features and Benefits
  • The click-stop system enables easy volume adjustment.
  • Broad volume display for better reading.
  • Color-coded for easy matching of fitting tips
  • High chemical and UV-resistance materials to ensure a long lifetime of the pipette.
  • Fully autoclavable and requires no disassembly.
  • Easy cleaning and maintenance with only three parts to disassemble.
  • Safe-cone filters provided for models >10μL.
  • Εrgοnomic finger support reduces the grip force required to hold the pipette.
  • Range of volume-3μL (5μL for fixed) to 10mL
  • Low pipetting forces
  • Wide range of adjustable single and multi-channel models and fixed single-channel models
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)