Skip to Content
Merck
All Photos(1)

Key Documents

Z707414

ThinSeal film

sterile

Synonym(s):

multiplate sealing film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22
Pricing and availability is not currently available.

sterility

sterile

feature

piercable: no

packaging

pack of 100 ea

manufacturer/tradename

Excel Scientific STR-THIN-PLT

technique(s)

ELISA: suitable

L

141 mm

W × L

5.4 cm × 14.1 cm

W

54 mm

Looking for similar products? Visit Product Comparison Guide

General description

Semi-pierceable, and leaves minimal residue. Polyester films just 25 μm thick, clearer and marginally pierceable with single-channel pipettes for sample recovery. Dimensions are 54.0 mm x 141.0 mm.

Legal Information

ThinSeal is a trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service