Key Documents
Z688193
HandEvac handheld aspirator
8-Channel plastic adapter with ejector for disposable tips
Synonym(s):
Handheld Vacuum Aspirator, Portable Aspirator
About This Item
UNSPSC Code:
41105100
NACRES:
NB.22
Pricing and availability is not currently available.
description
Adapter Only (hand operator not included)
manufacturer/tradename
Cole Parmer 1305037
General description
Rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes!
This ergonomic, hand-held aspirator can be used with pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications. A pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied. This prevents the accidental aspiration of cells or supernatant associated with normal vacuum techniques.
Use with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems. Autoclavable unit is supplied with a single-channel adapter for disposable pipette tips.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Questions
1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?
  2. What size tip needs to be used with the HandEvac handheld aspirator, Product Z688193?
    1 answer
    1. The HandEvac handheld aspirator, product Z688193, can be used with 200 μL pipette tips.
      Helpful?