Skip to Content
Merck
All Photos(1)

Key Documents

Z653381

Hettich® ROTOFIX 32A/Universal 320/320R centrifuge accessories

Adapter for 4-7 mL tubes 13x100 mm, for ROTOFIX 32 and UNIVERSAL 320

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.22

feature

Holds 5 / 6 / 7 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Hettich 1383

technique(s)

centrifugation: suitable

compatibility

for use with 1481, 1490, 1495 (Rotors)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service