Skip to Content
Merck
All Photos(1)

Key Documents

Z377821

Advanced PAP Pen

5 mm tip width

Synonym(s):

hydrophobic barrier pen, immunostaining wax pen, liquid blocker pap pen, water repellant barrier pen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41181600
NACRES:
MA.01

packaging

pack of 1 ea

manufacturer/tradename

Ted Pella 22309

Looking for similar products? Visit Product Comparison Guide

Application

Use pen to draw a hydrophobic circle around slide-mounted tissue. Prevents the waste of valuable reagents by keeping liquid pooled in a small droplet. Insoluble in ethanol, acetone and water, but will wash off with xylene.

For use with paraffin-mounted sections or slides treated with albumin or poly-lysine.

Quantity

Each pen can make more than 600 circles.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Ozone 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Liver,Central nervous system, Respiratory system

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mimi R Borrelli et al.
Aesthetic surgery journal, 40(4), 369-379 (2019-04-12)
Breast capsular contracture is a major problem following implant-based breast reconstruction, particularly in the setting of radiation therapy. Recent work has identified a fibrogenic fibroblast subpopulation characterized by CD26 surface marker expression. This work aimed to investigate the role of
Bruce A Corliss et al.
Frontiers in physiology, 10, 1389-1389 (2019-11-30)
Frigid temperatures of the Southern Ocean are known to be an evolutionary driver in Antarctic fish. For example, many fish have reduced red blood cell (RBC) concentration to minimize vascular resistance. Via the oxygen-carrying protein hemoglobin, RBCs contain the vast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service