Z373206
Plant tissue culture system temporary immersion
pore size 1 μm, filter diam. 37 mm
About This Item
Recommended Products
filter diam.
37 mm
pore size
1 μm
application(s)
agriculture
Related Categories
General description
The air flow oxygenates and agitates the medium. When the flow is turned off, pressure drops, and the medium returns to the bottom of the vessel. All components are autoclavable and reusable.
Using manifold and solenoid valves, one air pump can operate several vessels automatically. Manifold, air pump, inlet and outlet filters are not included.
The RITA® temporary immersion system for in vitro plant culture ensures improved yields compared with culture on a gel medium.
- Simple plant and medium handling
- Improved nutrition: the plant is in close contact with the medium during the immersion process
- The plant is covered by a capillary film (Polysulfone) throughout the remaining period of its immersion
- Marked reduction in asphyxiation
- Complete renewal of the culture atmosphere on each immersion
- Control of morphological processes by modifying immersion frequency and duration
- The two Inlet and Outlet filters have a pore size of 1 μm and are 37 mm in diameter
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service