Z326615
BRAND® BLAUBRAND® volumetric flask, PP stopper, clear glass
volume 200 mL, accuracy: 0.15 mL, polypropylene stopper, neck joint: ST/NS 14/23
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
polypropylene stopper
Joint
joint: ST/NS neck14/23
description
calibrated to contain (TC, IN)
sterility
non-sterile
feature
accuracy class A (DIN EN ISO 1042, calibrated to contain (TC, EX))
closure type PP Stopper
packaging
pack of 1
manufacturer/tradename
BRAND
volume measuring range
200 mL, accuracy: 0.15 mL
capacity
200 mL
General description
Volumetric flasks according to DIN EN ISO 1042, Class A, conformity certified. Made from Boro 3.3. Supplied with a square-grip PP stopper. Marks and inscriptions in high contast blue enamel.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
BR37250 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG