Skip to Content
Merck
All Photos(1)

Key Documents

VIV102016

SynVivo SynTox 3D Toxicology model chip, IMN2 radial configuration

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NB.35

packaging

case of 1 ea

manufacturer/tradename

SynVivo 102016

storage temp.

room temp

General description

SynTox 3D toxicology model chip- Barrier type: Slits, Slit width: 2 μm, Distance between slits: 50 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 μm. This chip is used to create the SynTox model in monoculture, co or tri-culture configurations.

SynTox is the only commercially available 3D toxicology model with real-time optical monitoring and multi-compartment, multi-cellular architecture and low reagent requirements. Other benefits of this platform are:

  • Physiologically realistic morphological, fluidic and 3D cellular conditions
  • Universal platform with architecture specific of desired organ
  • Significant reduction in cost and time
  • Robust and easy to use protocols
  • Compatible with standard analytical instruments for both on chip and off chip assays including omic methodologies for systems biology and bioinformatic analysis

Linkage

LInk to Publications
application text

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service