Skip to Content
Merck
All Photos(1)

Key Documents

SML3901

Sigma-Aldrich

Crenolanib trifluoroacetate

≥98% (HPLC)

Synonym(s):

1-[2-[5-[(3-Methyloxetan-3-yl)methoxy]benzimidazol-1-yl]quinolin-8-yl]piperidin-4-amine trifluoroacetate, 1-[2-[5-[(3-methyl-3-oxetanyl)methoxy]-1H-benzimidazol-1-yl]-8-quinolinyl]-4-piperidinamine trifluoroacetate, ARO 00 trifluoroacetate2, CP 868596 trifluoroacetate, CP868596 trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29N5O2 · xC2HF3O2
Molecular Weight:
443.54 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N6(CCC(CC6)N)c1c2nc(ccc2ccc1)[n]3c4c(nc3)cc(cc4)OCC5(COC5)C

InChI

1S/C26H29N5O2/c1-26(14-32-15-26)16-33-20-6-7-22-21(13-20)28-17-31(22)24-8-5-18-3-2-4-23(25(18)29-24)30-11-9-19(27)10-12-30/h2-8,13,17,19H,9-12,14-16,27H2,1H3

InChI key

DYNHJHQFHQTFTP-UHFFFAOYSA-N

Biochem/physiol Actions

Crenolanib is an orally bioavailable, potent and selective platelet-derived growth factor receptor (PDGFR) tyrosine kinase inhibitor. Also crenolanib potently inhibits FLT3.
Platelet-derived growth factor receptor (PDGFR) tyrosine kinase inhibitor

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activation of PDGFRA signaling contributes to filamin C-related arrhythmogenic cardiomyopathy
Science Advances, 8(8), eabk0052-eabk0052 (2022)
Phase I study of the safety, tolerability, and pharmacokinetics of oral CP-868,596, a highly specific platelet-derived growth factor receptor tyrosine kinase inhibitor in patients with advanced cancers
Journal of Clinical Oncology, 27(31), 5262-5269 (2009)
Crenolanib inhibits the drug-resistant PDGFRA D842V mutation associated with imatinib-resistant gastrointestinal stromal tumors
Clinical Cancer Research, 18(16), 4375-4384 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service