Skip to Content
Merck
All Photos(1)

Key Documents

SML3891

Sigma-Aldrich

Letermovir

≥98% (HPLC)

Synonym(s):

(4S)-2-{8-Fluoro-2-[4-(3-methoxyphenyl)piperazin-1-yl]-3-[2-methoxy-5-(trifluoromethyl)phenyl]-3,4-dihydroquinazolin-4-yl}acetic acid, AIC 246, AIC-246, AIC246

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H28F4N4O4
CAS Number:
Molecular Weight:
572.55
MDL number:
UNSPSC Code:
41106609
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(c(cc1)OC)N2[C@H](c3c(c(ccc3)F)N=C2N4CCN(CC4)c5cc(ccc5)OC)CC(=O)O

InChI

1S/C29H28F4N4O4/c1-40-20-6-3-5-19(16-20)35-11-13-36(14-12-35)28-34-27-21(7-4-8-22(27)30)23(17-26(38)39)37(28)24-15-18(29(31,32)33)9-10-25(24)41-2/h3-10,15-16,23H,11-14,17H2,1-2H3,(H,38,39)/t23-/m0/s1

InChI key

FWYSMLBETOMXAG-QHCPKHFHSA-N

Biochem/physiol Actions

Letermovir (AIC246) is an orally active, potent and selective human cytomegalovirus (HCMV) DNA terminase complex inhibitor that blocks HCMV replication (EC50 = 0.9-3.1 nM among 17 clinial HCMV isolates; EC50 = 1.6-3.9 nM against 6 AD169-derived drug-resistant variants) by interfering with viral DNA cleavage/packaging. Letermovir exhibits little potency toward MCMV (Smith), RCMV (Maastricht), VZV (Oka), HSV-1 (166v VP22-GFP), HSV-2 (01-6332), HHV-6 (typeA-GS) or EBV (B95-8) (MCMV EC50 = 4.5 µM, all other EC50 >10 µM).

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 Oral

Target Organs

Liver,Blood,spleen

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro and in vivo activities of the novel anticytomegalovirus compound AIC246
Antimicrobial Agents and Chemotherapy, 54(3), 1290-1297 (2010)
In vitro evaluation of the activities of the novel anticytomegalovirus compound AIC246 (letermovir) against herpesviruses and other human pathogenic viruses
Antimicrobial Agents and Chemotherapy, 56(2), 1135-1137 (2012)
The novel anticytomegalovirus compound AIC246 (Letermovir) inhibits human cytomegalovirus replication through a specific antiviral mechanism that involves the viral terminase
Journal of virology, 85(20), 10884-10893 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service