Skip to Content
Merck
All Photos(2)

Key Documents

SML3648

Sigma-Aldrich

Zuclomiphene

≥98% (HPLC)

Synonym(s):

(Z)- 2-[4-(2-Chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethyl-ethanamine, (Z)-Clomiphene, 2-[4-[(1Z)-2-Chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine, cis-Clomifene, cis-Clomiphene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28ClNO
CAS Number:
Molecular Weight:
405.96
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Orally available, less active cis isomer of the fertility drug clomiphene; mixed agonist of estrogen receptor.Zuclomiphene is an orally available, less active cis isomer of the fertility drug clomiphene. Zuclomiphene is a mixed agonist of estrogen receptor responsible for the estrogenic activity of clomiphene and therefore the common side effects sometimes seen in males. Similarly, to clomiphene and enclomiphene, zuclomiphene blocks Ebola virus entry and infection in tissue culture cells and affords up to 90% protection in the mouse model of Ebola virus disease.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - STOT RE 2 Oral

Target Organs

ovary

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypothalamic-Pituitary-Testicular Axis Effects and Urinary Detection Following Clomiphene Administration in Males
The Journal of Clinical Endocrinology and Metabolism, 104(3), 906-914 (2019)
Serum levels of enclomiphene and zuclomiphene in men with hypogonadism on long-term clomiphene citrate treatment
BJU International, 119(1), 171-176 (2017)
Clomiphene and Its Isomers Block Ebola Virus Particle Entry and Infection with Similar Potency: Potential Therapeutic Implications
Viruses, 8(8), 206-206 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service