SML3394
GZ-11608
≥98% (HPLC)
Synonym(s):
4-Methoxy-N-[(1S)-1-methyl-2-phenylethyl]-benzenepropanamine, S-3-(4-Methoxyphenyl)-N-(1-phenylpropan-2-yl)propan-1-amine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
oil
color
colorless to brown
storage temp.
2-8°C
SMILES string
N([C@H](Cc2ccccc2)C)CCCc1ccc(cc1)OC
InChI key
LKTLDVZCNLGHOG-INIZCTEOSA-N
Biochem/physiol Actions
GZ-11608 is a potent and selective inhibitor of vesicular monoamine transporter-2 (VMAT2) that diminished hERG interaction. GZ-11608 reduces behavioral effects of methamphetamine such as locomotor activity, self-administration, and methamphetamine seeking behavior. It does not exhibit abuse liability. GZ-11608 inhibits methamphetamine-induced dopamine release from isolated synaptic vesicles from brain dopaminergic neurons.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of pharmacology and experimental therapeutics, 371(2), 526-543 (2019-08-16)
Despite escalating methamphetamine use and high relapse rates, pharmacotherapeutics for methamphetamine use disorders are not available. Our iterative drug discovery program had found that R-N-(1,2-dihydroxypropyl)-2,6-cis-di-(4-methoxyphenethyl)piperidine hydrochloride (GZ-793A), a selective vesicular monoamine transporter-2 (VMAT2) inhibitor, specifically decreased methamphetamine's behavioral effects. However
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service