Skip to Content
Merck
All Photos(1)

Key Documents

SML3117

Sigma-Aldrich

N-Desethyl sunitinib hydrochloride

≥98% (HPLC)

Synonym(s):

5-(5-Fluoro-2-oxo-1,2-dihydroindol-(3Z)-ylidenemethyl)-2,4-dimethyl-1H-pyrrole-3-carboxylic acid (2-ethylaminoethyl)amide hydrochloride, N-Desethylsunitinib hydrochloride, N-[2-(Ethylamino)ethyl]-5-[(Z)-(5-fluoro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxamide hydrochloride, SU 12662 hydrochloride, SU12662 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23FN4O2 · xHCl
CAS Number:
Molecular Weight:
370.42 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Fc1cc2c(cc1)NC(=O)\C\2=C/c3[nH]c(c(c3C)C(=O)NCCNCC)C

InChI

1S/C20H23FN4O2/c1-4-22-7-8-23-20(27)18-11(2)17(24-12(18)3)10-15-14-9-13(21)5-6-16(14)25-19(15)26/h5-6,9-10,22,24H,4,7-8H2,1-3H3,(H,23,27)(H,25,26)/b15-10-

InChI key

LIZNIAKSBJKPQC-GDNBJRDFSA-N

Biochem/physiol Actions

N-Desethyl sunitinib is a multi-target tyrosine kinase inhibitor that is a major active metabolite of an anticancer drug sunitinib. N-Desethyl sunitinib has a similar kinase inhibitory effect as sunitinib. In patients, plasma sunitinib and N-desethyl sunitinib accounts for 42% and 24%, respectively. Thus N-desethyl sunitinib accounts for one third of the therapeutic effect of oral sunitinib.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2

Target Organs

blood vessel,Bone marrow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service